Product Details
- SNP ID
-
rs148087489
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:35619583 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCTCTTCAGCCTGCAGTGGGACC[C/G]GCGCGTCATGCAGCGTGCCACCAGC
- Phenotype
-
MIM: 612206
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FJX1
PubMed Links
Gene Details
- Gene
- FJX1
- Gene Name
- four jointed box 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014344.3 |
1397 |
Missense Mutation |
CCG,CGG |
P316R |
NP_055159.2 |
View Full Product Details