Product Details

SNP ID
rs148118632
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118528385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCTCTTGGCAATGGGGAACCA[A/G]CCAAGGCCCCTTCCCAGGTCTCTTG
Phenotype
MIM: 159555 MIM: 613934
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KMT2A PubMed Links

Gene Details

Gene
KMT2A
Gene Name
lysine methyltransferase 2A
There are no transcripts associated with this gene.

Gene
LOC101929089
Gene Name
uncharacterized LOC101929089
There are no transcripts associated with this gene.

Gene
TMEM25
Gene Name
transmembrane protein 25
There are no transcripts associated with this gene.

Gene
TTC36
Gene Name
tetratricopeptide repeat domain 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080441.1 Intron NP_001073910.1

View Full Product Details