Product Details

SNP ID
rs148141652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:96391991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTATGAAGTGTTAACTGATAGC[A/G]AAATCATCAGAAGAGCACAAGGCCA
Phenotype
MIM: 603211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC82 PubMed Links

Gene Details

Gene
CCDC82
Gene Name
coiled-coil domain containing 82
There are no transcripts associated with this gene.

Gene
JRKL
Gene Name
JRK-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261833.1 1998 Missense Mutation AAA,GAA K448E NP_001248762.1
NM_003772.3 1998 Missense Mutation AAA,GAA K448E NP_003763.2

View Full Product Details