Product Details

SNP ID
rs148346044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126271475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCTGAGATTAAGAAGAAGATC[A/C]AGTCGATCCTGCCTGGAAGGTCCTG
Phenotype
MIM: 616587 MIM: 613622 MIM: 182180 MIM: 606252
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017547.3 1423 Missense Mutation AAG,CAG K42Q NP_060017.1
XM_006718879.3 1423 Intron XP_006718942.1
XM_006718881.3 1423 Intron XP_006718944.1
XM_011542895.2 1423 UTR 5 XP_011541197.1
XM_011542896.2 1423 UTR 5 XP_011541198.1
XM_017018000.1 1423 Missense Mutation AAG,CAG K42Q XP_016873489.1
XM_017018001.1 1423 UTR 5 XP_016873490.1
XM_017018002.1 1423 Intron XP_016873491.1
XM_017018003.1 1423 UTR 5 XP_016873492.1
XM_017018004.1 1423 UTR 5 XP_016873493.1
XM_017018005.1 1423 UTR 5 XP_016873494.1
XM_017018006.1 1423 UTR 5 XP_016873495.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details