Product Details

SNP ID
rs148394356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72228369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGACCTCGCAGGGTGTGCCTGTG[C/T]GCCGCTTCCAGACCCTGGGTGAGCT
Phenotype
MIM: 136425 MIM: 600829
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOLR2 PubMed Links

Gene Details

Gene
FOLR2
Gene Name
folate receptor beta
There are no transcripts associated with this gene.

Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001567.3 573 Missense Mutation CGC,TGC R90C NP_001558.3
XM_005273978.3 573 Missense Mutation CGC,TGC R90C XP_005274035.1
XM_005273979.4 573 Missense Mutation CGC,TGC R90C XP_005274036.1
XM_011544999.2 573 Missense Mutation CGC,TGC R90C XP_011543301.1
XM_011545000.2 573 Missense Mutation CGC,TGC R90C XP_011543302.1
XM_017017673.1 573 Missense Mutation CGC,TGC R80C XP_016873162.1

View Full Product Details