Product Details

SNP ID
rs148446106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6682157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAGCTGTGTGGGAGCTGTGGTT[C/G]CTTGCTTCCTGGCTTTGCCTGAGGT
Phenotype
MIM: 611830
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL17 PubMed Links

Gene Details

Gene
MRPL17
Gene Name
mitochondrial ribosomal protein L17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022061.3 594 Missense Mutation AGC,AGG S163R NP_071344.1

View Full Product Details