Product Details

SNP ID
rs148465141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55572281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATTAGTGTCAAGACCAGCCAG[A/G]CACTTAAGAACCCAATGTTCTTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C16 PubMed Links

Gene Details

Gene
OR4C16
Gene Name
olfactory receptor family 4 subfamily C member 16 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004701.2 154 Missense Mutation ACA,GCA T52A NP_001004701.2

View Full Product Details