Product Details

SNP ID
rs148475449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1697623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGTGTCCCCATTTGCTGCCA[A/G]TGCAAAATCTGAGGCTCTGACTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP5-6 PubMed Links

Gene Details

Gene
KRTAP5-6
Gene Name
keratin associated protein 5-6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012416.1 429 Silent Mutation CAA,CAG Q126Q NP_001012416.1

View Full Product Details