Product Details

SNP ID
rs148672786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55774126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATTTTCACTACCATTATGAAGA[C/T]GCGATCTGCAAGTGGGCGCCAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5D13 PubMed Links

Gene Details

Gene
OR5D13
Gene Name
olfactory receptor family 5 subfamily D member 13 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001967.1 689 Missense Mutation ACG,ATG T230M NP_001001967.1

View Full Product Details