Product Details

SNP ID
rs148729565
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27367940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTACTCTGGTAGAAGCAGGCT[C/T]GTCCACAGGCCTGCACCTGGTCAGA
Phenotype
MIM: 612324 MIM: 606666
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC34 PubMed Links

Gene Details

Gene
CCDC34
Gene Name
coiled-coil domain containing 34
There are no transcripts associated with this gene.

Gene
LGR4
Gene Name
leucine rich repeat containing G protein-coupled receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018490.2 3268 Missense Mutation CAA,CGA Q928R NP_060960.2

View Full Product Details