Product Details

SNP ID
rs148734778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:210595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGACGCTGCCCTTTACCGACAC[C/T]TGGGGACCCTTCTCCGGCACTGTGT
Phenotype
MIM: 615417 MIM: 609146 MIM: 604481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BET1L PubMed Links

Gene Details

Gene
BET1L
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
There are no transcripts associated with this gene.

Gene
MIR6743
Gene Name
microRNA 6743
There are no transcripts associated with this gene.

Gene
RIC8A
Gene Name
RIC8 guanine nucleotide exchange factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286134.1 1281 Silent Mutation CTG,TTG L251L NP_001273063.1
NM_021932.5 1281 Silent Mutation CTG,TTG L251L NP_068751.4
XM_005253052.3 1281 Silent Mutation CTG,TTG L255L XP_005253109.1
XM_017018107.1 1281 Silent Mutation CTG,TTG L141L XP_016873596.1
Gene
SIRT3
Gene Name
sirtuin 3
There are no transcripts associated with this gene.

View Full Product Details