Product Details

SNP ID
rs148751060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:88507860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATCTTCCACTGGTTTTCTTGG[A/T]TAATGGAGGTGAATCTGAAATAAGT
Phenotype
MIM: 604102
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GRM5 PubMed Links

Gene Details

Gene
GRM5
Gene Name
glutamate metabotropic receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000842.4 4767 UTR 3 NP_000833.1
NM_001143831.2 4767 UTR 3 NP_001137303.1
XM_006718828.3 4767 UTR 3 XP_006718891.1
XM_011542792.1 4767 UTR 3 XP_011541094.1
XM_017017627.1 4767 UTR 3 XP_016873116.1
Gene
GRM5-AS1
Gene Name
GRM5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details