Product Details

SNP ID
rs148802656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65891243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGAATGGGGCCCCCGCGGCGGC[C/G]CCAGCGGCGCCGGGGGATCAGGAGC
Phenotype
MIM: 605360 MIM: 602364 MIM: 608296 MIM: 136515
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC85B PubMed Links

Gene Details

Gene
CCDC85B
Gene Name
coiled-coil domain containing 85B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006848.2 840 Missense Mutation CCC,GCC P154A NP_006839.2
Gene
CTSW
Gene Name
cathepsin W
There are no transcripts associated with this gene.

Gene
FIBP
Gene Name
FGF1 intracellular binding protein
There are no transcripts associated with this gene.

Gene
FOSL1
Gene Name
FOS like 1, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300844.1 840 Intron NP_001287773.1
NM_001300855.1 840 Intron NP_001287784.1
NM_001300856.1 840 Intron NP_001287785.1
NM_001300857.1 840 Intron NP_001287786.1
NM_005438.4 840 Intron NP_005429.1

View Full Product Details