Product Details

SNP ID
rs148851960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118998522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGGTGCCTGTGCTGCGGCTG[G/T]GAGGTGCGGGAACACCTGAGCCATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC84 PubMed Links

Gene Details

Gene
CCDC84
Gene Name
coiled-coil domain containing 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198489.2 278 Missense Mutation TGG,TGT W71C NP_940891.1
XM_011542799.1 278 UTR 5 XP_011541101.1
XM_011542800.2 278 Intron XP_011541102.1
XM_017017652.1 278 UTR 5 XP_016873141.1
Gene
RPL23AP64
Gene Name
ribosomal protein L23a pseudogene 64
There are no transcripts associated with this gene.

View Full Product Details