Product Details

SNP ID
rs148897398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5152178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGTGGCTCCTAGCATGCCTAA[C/G]AAAATGTACAAGGGCTCATGGAGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR52A1 PubMed Links

Gene Details

Gene
OR52A1
Gene Name
olfactory receptor family 52 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012375.2 192 Silent Mutation TTC,TTG F64L NP_036507.2

View Full Product Details