Product Details

SNP ID
rs148975274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68688918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTGAGTTTCTGTCTTTCTTGC[A/G]TCTCAAAGGTATGTGAAATATCATC
Phenotype
MIM: 137035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAL PubMed Links

Gene Details

Gene
GAL
Gene Name
galanin and GMAP prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015973.4 551 Missense Mutation CAT,CGT H98R NP_057057.2

View Full Product Details