Product Details

SNP ID
rs148991955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58118745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACATGTGGAGAAAGTCTTGGCCC[C/T]GCCACCTGAAGACTTCACTTTCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9I1 PubMed Links

Gene Details

Gene
OR9I1
Gene Name
olfactory receptor family 9 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005211.1 700 Missense Mutation AGG,GGG R234G NP_001005211.1
Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 700 Intron NP_001005212.1

View Full Product Details