Product Details

SNP ID
rs148993599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61803285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAATGCTGTTTGGGGGACTTT[C/T]TGTTTTTGCTGTTTTCACCTACGCA
Phenotype
MIM: 606148 MIM: 600393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013402.4 Intron NP_037534.3
XM_011545022.2 Intron XP_011543324.1
Gene
FEN1
Gene Name
flap structure-specific endonuclease 1
There are no transcripts associated with this gene.

View Full Product Details