Product Details

SNP ID
rs149043465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62210458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACATGGTTGAAAAGACCATCA[A/G]TTCCGACATATCTATACCTGAATAC
Phenotype
MIM: 604398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCGB2A1 PubMed Links

Gene Details

Gene
SCGB2A1
Gene Name
secretoglobin family 2A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002407.2 165 Missense Mutation AAT,AGT N34S NP_002398.1

View Full Product Details