Product Details

SNP ID
rs149058797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65045883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGGTCCGTTCCAAGAGCATCA[A/G]CTGGTCATTGAGCATCCGGACCTGC
Phenotype
MIM: 602640 MIM: 605964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL2-SNX15 PubMed Links

Gene Details

Gene
ARL2-SNX15
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
NAALADL1
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005468.2 1991 Silent Mutation CTG,TTG L659L NP_005459.2
XM_011544706.2 1991 Silent Mutation CTG,TTG L656L XP_011543008.1
XM_011544707.2 1991 Silent Mutation CTG,TTG L611L XP_011543009.1
XM_011544708.2 1991 Silent Mutation CTG,TTG L606L XP_011543010.1
XM_011544709.2 1991 Intron XP_011543011.1
XM_011544710.2 1991 Intron XP_011543012.1
XM_011544711.2 1991 Intron XP_011543013.1
XM_011544712.2 1991 Intron XP_011543014.1
Gene
SAC3D1
Gene Name
SAC3 domain containing 1
There are no transcripts associated with this gene.

Gene
SNX15
Gene Name
sorting nexin 15
There are no transcripts associated with this gene.

View Full Product Details