Product Details

SNP ID
rs149064670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6318948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCCTTCTCACGGAGTCTGGTT[A/C]GGGTGAAATTACTTTTATTGATGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 1035 UTR 3 NP_659477.2

View Full Product Details