Product Details
- SNP ID
-
rs149241583
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:67405649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGATGGAGACCATTGGCAGGGAC[C/T]TGCACCGTCAATTCCCTCTGCACGA
- Phenotype
-
MIM: 176875
MIM: 603761
MIM: 610831
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP1CA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12805811] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP1CA
- Gene Name
- protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.
- Gene
- RAD9A
- Gene Name
- RAD9 checkpoint clamp component A
There are no transcripts associated with this gene.
- Gene
- TBC1D10C
- Gene Name
- TBC1 domain family member 10C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256508.1 |
731 |
Silent Mutation |
CTG,TTG |
L139L |
NP_001243437.1 |
NM_198517.3 |
731 |
Silent Mutation |
CTG,TTG |
L139L |
NP_940919.1 |
XM_006718538.3 |
731 |
Silent Mutation |
CTG,TTG |
L139L |
XP_006718601.1 |
XM_006718539.3 |
731 |
Silent Mutation |
CTG,TTG |
L139L |
XP_006718602.1 |
XM_006718540.3 |
731 |
Silent Mutation |
CTG,TTG |
L139L |
XP_006718603.1 |
XM_006718541.3 |
731 |
Missense Mutation |
CCT,CTT |
P39L |
XP_006718604.1 |
XM_006718542.3 |
731 |
Missense Mutation |
CCT,CTT |
P39L |
XP_006718605.1 |
XM_006718543.2 |
731 |
Missense Mutation |
CCT,CTT |
P39L |
XP_006718606.1 |
XM_006718544.3 |
731 |
Missense Mutation |
CCT,CTT |
P39L |
XP_006718607.1 |
XM_011545001.2 |
731 |
Missense Mutation |
CCT,CTT |
P39L |
XP_011543303.1 |
XM_011545002.2 |
731 |
Silent Mutation |
CTG,TTG |
L139L |
XP_011543304.1 |
View Full Product Details