Product Details

SNP ID
rs149354945
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35432638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATGCTGCTCCTCACACAGCAG[C/T]GAGTCCACCACACTGACCACCCCAG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PAMR1 PubMed Links

Gene Details

Gene
PAMR1
Gene Name
peptidase domain containing associated with muscle regeneration 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001991.2 2378 Silent Mutation TCA,TCG S627S NP_001001991.1
NM_001282675.1 2378 Silent Mutation TCA,TCG S587S NP_001269604.1
NM_001282676.1 2378 Silent Mutation TCA,TCG S516S NP_001269605.1
NM_015430.3 2378 Silent Mutation TCA,TCG S644S NP_056245.2

View Full Product Details