Product Details

SNP ID
rs149409172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60790510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAAGAAGAGCAGCCTTCTTAAG[A/G]AGCTGGGGGTGAGCATCCACTTCCC
Phenotype
MIM: 608403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A10 PubMed Links

Gene Details

Gene
MS4A10
Gene Name
membrane spanning 4-domains A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206893.3 271 Missense Mutation AAG,GAG K59E NP_996776.2
XM_011544989.1 271 Missense Mutation AAG,GAG K59E XP_011543291.1
XM_011544991.2 271 Intron XP_011543293.1

View Full Product Details