Product Details

SNP ID
rs149436433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102772943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTATGGTTGGAGGAAAACCCAGG[A/G]TATGGATGCCTCTTGGATAACCTGC
Phenotype
MIM: 185260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP10 PubMed Links

Gene Details

Gene
MMP10
Gene Name
matrix metallopeptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002425.2 1167 Missense Mutation ACC,ATC T377I NP_002416.1

View Full Product Details