Product Details

SNP ID
rs149487173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119639883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCACCCGGCCCCATCCGTCTCC[A/G]GTGGGCTCTTCTGCTGCCGGTTGTA
Phenotype
MIM: 600644
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NECTIN1 PubMed Links

Gene Details

Gene
NECTIN1
Gene Name
nectin cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002855.4 1305 Intron NP_002846.3
NM_203285.1 1305 Missense Mutation NP_976030.1
NM_203286.1 1305 Intron NP_976031.1

View Full Product Details