Product Details

SNP ID
rs149495312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55554712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGTCATCACCCCAAAGATGATT[A/G]TAGACTCCCTCTATGTGACAAAAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C15 PubMed Links

Gene Details

Gene
OR4C15
Gene Name
olfactory receptor family 4 subfamily C member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001920.1 406 Missense Mutation ATA,GTA I136V NP_001001920.1

View Full Product Details