Product Details

SNP ID
rs149718333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65356403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATGTGATCAAGTACAGGTGCTG[C/T]TGTGTGCCAACAGCAGGGGCACCGA
Phenotype
MIM: 601671
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPF2 PubMed Links

Gene Details

Gene
DPF2
Gene Name
double PHD fingers 2
There are no transcripts associated with this gene.

Gene
TIGD3
Gene Name
tigger transposable element derived 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145719.2 758 Silent Mutation CTG,TTG L199L NP_663771.1

View Full Product Details