Product Details

SNP ID
rs149726248
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62625819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCCAGAACATCCCTTGGGTTAT[C/G]GCAGGTGAATACTCCAATCAGATGC
Phenotype
MIM: 606374 MIM: 104160
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
B3GAT3 PubMed Links

Gene Details

Gene
B3GAT3
Gene Name
beta-1,3-glucuronyltransferase 3
There are no transcripts associated with this gene.

Gene
GANAB
Gene Name
glucosidase II alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278192.1 3024 Missense Mutation CCA,CGA P852R NP_001265121.1
NM_001278193.1 3024 Missense Mutation CCA,CGA P830R NP_001265122.1
NM_001278194.1 3024 Missense Mutation CCA,CGA P847R NP_001265123.1
NM_198334.2 3024 Missense Mutation CCA,CGA P944R NP_938148.1
NM_198335.3 3024 Missense Mutation CCA,CGA P966R NP_938149.2
XM_017017412.1 3024 Missense Mutation CCA,CGA P847R XP_016872901.1
XM_017017413.1 3024 Missense Mutation CCA,CGA P703R XP_016872902.1
XM_017017414.1 3024 Missense Mutation CCA,CGA P703R XP_016872903.1

View Full Product Details