Product Details

SNP ID
rs150209910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5515420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTGTCATTGGCTGAGGTGTTT[A/G]AAGAGAAACCACCAGAGCTATTATA
Phenotype
MIM: 605473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFM5P PubMed Links

Gene Details

Gene
OLFM5P
Gene Name
olfactomedin family member 5, pseudogene
There are no transcripts associated with this gene.

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
There are no transcripts associated with this gene.

Gene
UBQLN3
Gene Name
ubiquilin 3
There are no transcripts associated with this gene.

Gene
UBQLNL
Gene Name
ubiquilin-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145053.4 1307 Missense Mutation TCA,TTA S341L NP_659490.4

View Full Product Details