Product Details

SNP ID
rs150220771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:44067769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGACCAGAAGCTGCCAGAGCTC[C/T]GTGGAGTGGGTGATCCTGCCATGAT
Phenotype
MIM: 608405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACCS PubMed Links

Gene Details

Gene
ACCS
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127219.1 729 Missense Mutation CGT,TGT R48C NP_001120691.1
NM_032592.3 729 Missense Mutation CGT,TGT R48C NP_115981.1
XM_005253170.3 729 Missense Mutation CGT,TGT R149C XP_005253227.2
XM_005253172.2 729 Intron XP_005253229.1
XM_006718348.2 729 Intron XP_006718411.1
XM_011520406.2 729 Missense Mutation CGT,TGT R149C XP_011518708.1
XM_011520407.1 729 Intron XP_011518709.1
XM_011520410.2 729 UTR 5 XP_011518712.1
XM_017018421.1 729 Missense Mutation CGT,TGT R149C XP_016873910.1
Gene
ACCSL
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
There are no transcripts associated with this gene.

View Full Product Details