Product Details

SNP ID
rs150413720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72109463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCAGGGATGCCCTGGCCTTC[C/T]CTAACTCTGGCCCACCTAGCCAATT
Phenotype
MIM: 614717 MIM: 613510 MIM: 612414
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC15 PubMed Links

Gene Details

Gene
ANAPC15
Gene Name
anaphase promoting complex subunit 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278485.1 2290 Intron NP_001265414.1
NM_001278486.1 2290 Intron NP_001265415.1
NM_001278487.1 2290 Intron NP_001265416.1
NM_001278488.1 2290 Intron NP_001265417.1
NM_001278489.1 2290 Intron NP_001265418.1
NM_001278490.1 2290 Intron NP_001265419.1
NM_001278491.1 2290 Intron NP_001265420.1
NM_001278492.1 2290 Intron NP_001265421.1
NM_001278493.1 2290 Intron NP_001265422.1
NM_001278494.1 2290 Intron NP_001265423.1
NM_014042.2 2290 Intron NP_054761.1
XM_017017488.1 2290 Intron XP_016872977.1
XM_017017489.1 2290 Intron XP_016872978.1
XM_017017490.1 2290 Intron XP_016872979.1
XM_017017491.1 2290 Intron XP_016872980.1
XM_017017492.1 2290 Intron XP_016872981.1
XM_017017493.1 2290 Intron XP_016872982.1
XM_017017494.1 2290 Intron XP_016872983.1
XM_017017495.1 2290 Intron XP_016872984.1
XM_017017496.1 2290 Intron XP_016872985.1
XM_017017497.1 2290 Intron XP_016872986.1
Gene
LAMTOR1
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
There are no transcripts associated with this gene.

Gene
LRTOMT
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145307.4 2290 Intron NP_001138779.1
NM_001145308.4 2290 UTR 3 NP_001138780.1
NM_001145309.3 2290 UTR 3 NP_001138781.1
NM_001145310.3 2290 UTR 3 NP_001138782.1
NM_001205138.3 2290 Intron NP_001192067.1
NM_001271471.2 2290 Intron NP_001258400.1
NM_001318803.1 2290 Intron NP_001305732.1
NM_145309.5 2290 Intron NP_660352.1
XM_006718473.3 2290 Intron XP_006718536.1
XM_006718474.3 2290 Intron XP_006718537.1
XM_011544847.2 2290 Intron XP_011543149.1
XM_011544848.2 2290 Intron XP_011543150.1
XM_017017356.1 2290 Intron XP_016872845.1
XM_017017357.1 2290 UTR 3 XP_016872846.1
XM_017017358.1 2290 Intron XP_016872847.1
XM_017017359.1 2290 Intron XP_016872848.1
XM_017017360.1 2290 Intron XP_016872849.1

View Full Product Details