Product Details

SNP ID
rs150513236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56470414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGATGATGTAGACCACAAGAAA[A/G]ATGATGAAGAAGAGAACTTGCCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5M3 PubMed Links

Gene Details

Gene
OR5M3
Gene Name
olfactory receptor family 5 subfamily M member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004742.2 Intron NP_001004742.2
Gene
OR5M9
Gene Name
olfactory receptor family 5 subfamily M member 9
There are no transcripts associated with this gene.

View Full Product Details