Product Details

SNP ID
rs150529970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3856283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACCTAGAGTCATGGATGTATGC[A/G]TCCGTCTTGCCCTGTGGCTCCTCTG
Phenotype
MIM: 605921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4687 PubMed Links

Gene Details

Gene
MIR4687
Gene Name
microRNA 4687
There are no transcripts associated with this gene.

Gene
STIM1
Gene Name
stromal interaction molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277961.1 581 Missense Mutation ATC,GTC I5V NP_001264890.1
NM_001277962.1 581 Missense Mutation ATC,GTC I5V NP_001264891.1
NM_003156.3 581 Missense Mutation ATC,GTC I5V NP_003147.2

View Full Product Details