Product Details

SNP ID
rs150739333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6319260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGAGGCTCTGGCTCCAGCGTG[A/G]GCTCCAGCGCAGGCTGGGCTTCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 723 Missense Mutation CCC,CTC P230L NP_659477.2

View Full Product Details