Product Details
- SNP ID
-
rs150915961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74994613 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCCCAGCCCTGCTCTACATACCC[A/C]CCACCCACACCTTCCTGGCCTTTGC
- Phenotype
-
MIM: 604617
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NEU3
PubMed Links
Gene Details
- Gene
- NEU3
- Gene Name
- neuraminidase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006656.5 |
355 |
Missense Mutation |
ACC,CCC |
T67P |
NP_006647.3 |
View Full Product Details