Product Details
- SNP ID
-
rs151005116
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7987391 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTACCAGTAAGTGCTCCTCCGGC[C/T]ACGCCAACCCCAGTCCCGGCGGCGG
- Phenotype
-
MIM: 603914
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF3F
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61734576] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF3F
- Gene Name
- eukaryotic translation initiation factor 3 subunit F
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003754.2 |
|
Intron |
|
|
NP_003745.1 |
View Full Product Details