Product Details

SNP ID
rs151247449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:555874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGATGGACAAGTCGGTGCCGTC[A/G]GCGAAGACCCTCGGGGCCGGAGTCT
Phenotype
MIM: 143023
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMNTD2 PubMed Links

Gene Details

Gene
LMNTD2
Gene Name
lamin tail domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173573.2 1578 Silent Mutation GCC,GCT A478A NP_775844.2
XM_011519965.2 1578 Silent Mutation GCC,GCT A551A XP_011518267.2
XM_011519967.2 1578 Silent Mutation GCC,GCT A511A XP_011518269.2
XM_017017476.1 1578 Missense Mutation CCG,CTG P573L XP_016872965.1
XM_017017477.1 1578 Missense Mutation CCG,CTG P568L XP_016872966.1
XM_017017478.1 1578 Missense Mutation CCG,CTG P505L XP_016872967.1
XM_017017479.1 1578 Silent Mutation GCC,GCT A551A XP_016872968.1
Gene
LRRC56
Gene Name
leucine rich repeat containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198075.3 1578 Intron NP_932341.1
XM_005252775.2 1578 Intron XP_005252832.1
XM_005252776.2 1578 Intron XP_005252833.1
XM_006718132.2 1578 Intron XP_006718195.1
XM_006718133.2 1578 Intron XP_006718196.1
XM_011519875.2 1578 Intron XP_011518177.1
XM_011519876.1 1578 Intron XP_011518178.1
XM_011519877.2 1578 Intron XP_011518179.1
XM_017017167.1 1578 Intron XP_016872656.1
XM_017017168.1 1578 Intron XP_016872657.1
Gene
MIR210HG
Gene Name
MIR210 host gene
There are no transcripts associated with this gene.

Gene
RASSF7
Gene Name
Ras association domain family member 7
There are no transcripts associated with this gene.

View Full Product Details