Product Details

SNP ID
rs151269219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:88294184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATAGCCCACAAGCAGAACAGCA[C/T]GATTAGTCAGCTCAAAGGGGTTGAA
Phenotype
MIM: 602365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSC PubMed Links

Gene Details

Gene
CTSC
Gene Name
cathepsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114173.2 1329 Intron NP_001107645.1
NM_001814.5 1329 Missense Mutation CAT,CGT H405R NP_001805.3
NM_148170.4 1329 Intron NP_680475.1

View Full Product Details