Product Details

SNP ID
rs151271848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6319462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGCTCCTCGTCCGAGCTCTCTC[C/T]AACTTCGGCCTCCAGCTGCTCTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 521 Missense Mutation AGA,GGA R163G NP_659477.2

View Full Product Details