Product Details

SNP ID
rs137878349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26905232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTCGCAAAGAGAAGCCTAAG[A/C]ATTTTTTTAAAAACATTTCCAGAGA
Phenotype
MIM: 615079
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASUN PubMed Links

Gene Details

Gene
ASUN
Gene Name
asunder, spermatogenesis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018164.2 2456 UTR 3 NP_060634.2
XM_005253422.3 2456 UTR 3 XP_005253479.1
XM_011520750.2 2456 UTR 3 XP_011519052.1
XM_011520751.2 2456 UTR 3 XP_011519053.1
XM_011520752.2 2456 UTR 3 XP_011519054.1
XM_011520753.2 2456 UTR 3 XP_011519055.1
XM_017019632.1 2456 UTR 3 XP_016875121.1
XM_017019633.1 2456 UTR 3 XP_016875122.1
XM_017019634.1 2456 UTR 3 XP_016875123.1
XM_017019635.1 2456 UTR 3 XP_016875124.1
XM_017019636.1 2456 UTR 3 XP_016875125.1
XM_017019637.1 2456 UTR 3 XP_016875126.1

View Full Product Details