Product Details

SNP ID
rs137990615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96282573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACACATGTTTCATGGCCTCTTC[A/G]GCTGAAACCCTTTTCTTAGATTCAT
Phenotype
MIM: 603440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK17 PubMed Links

Gene Details

Gene
CDK17
Gene Name
cyclin dependent kinase 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170464.2 2024 Silent Mutation NP_001163935.1
NM_002595.4 2024 Silent Mutation NP_002586.2
XM_017019405.1 2024 Silent Mutation XP_016874894.1
XM_017019406.1 2024 Silent Mutation XP_016874895.1
XM_017019407.1 2024 Silent Mutation XP_016874896.1

View Full Product Details