Product Details

SNP ID
rs138059846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96909845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATGACATTGGTGGATAAATTCA[A/G]CCCACACACATCACCACATGGAATC
Phenotype
MIM: 600372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEDD1 PubMed Links

Gene Details

Gene
NEDD1
Gene Name
neural precursor cell expressed, developmentally down-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135175.1 425 Missense Mutation AAC,AGC N36S NP_001128647.1
NM_001135176.1 425 Missense Mutation AAC,AGC N29S NP_001128648.1
NM_001135177.1 425 Intron NP_001128649.1
NM_152905.3 425 Missense Mutation AAC,AGC N29S NP_690869.1
XM_005268644.2 425 Missense Mutation AAC,AGC N36S XP_005268701.1
XM_006719237.3 425 Intron XP_006719300.1
XM_011537903.2 425 Intron XP_011536205.1
XM_011537904.2 425 UTR 5 XP_011536206.1
XM_011537905.2 425 Intron XP_011536207.1
XM_011537906.2 425 Intron XP_011536208.1
XM_017018801.1 425 Intron XP_016874290.1
XM_017018802.1 425 Intron XP_016874291.1

View Full Product Details