Product Details

SNP ID
rs138068731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6639003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTGCTGGCTTTTGAAGGAACAG[A/T]AGTTTGGGTACTGGATATAGTCTGT
Phenotype
MIM: 608352 MIM: 606926
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACRBP PubMed Links

Gene Details

Gene
ACRBP
Gene Name
acrosin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032489.2 1508 Missense Mutation TAC,TTC Y487F NP_115878.2
Gene
LPAR5
Gene Name
lysophosphatidic acid receptor 5
There are no transcripts associated with this gene.

View Full Product Details