Product Details

SNP ID
rs138128077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56361706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCATTCTCACAGTCCTCTGTC[A/G]GGAGGGAGCGCCCGACCCTTCCTGT
Phenotype
MIM: 107760 MIM: 600556
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOF PubMed Links

Gene Details

Gene
APOF
Gene Name
apolipoprotein F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001638.2 554 Missense Mutation NP_001629.1
Gene
STAT2
Gene Name
signal transducer and activator of transcription 2
There are no transcripts associated with this gene.

View Full Product Details