Product Details

SNP ID
rs138194545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48472692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATGGGCAAATGGATTCATTTA[A/G]AGCTGCGGAACAGGACGCCCTCCGA
Phenotype
MIM: 606878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANP32D PubMed Links

Gene Details

Gene
ANP32D
Gene Name
acidic nuclear phosphoprotein 32 family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012404.2 Intron NP_036536.2
Gene
C12orf54
Gene Name
chromosome 12 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152319.3 Intron NP_689532.1
XM_005268636.3 Intron XP_005268693.1
XM_011537896.2 Intron XP_011536198.1
XM_017018796.1 Intron XP_016874285.1

View Full Product Details