Product Details

SNP ID
rs138261783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6329848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTTGGGGATGGGGTCGGAGGC[A/G]AGGGCTGTCGCAAGGATGGGGTCAG
Phenotype
MIM: 611743 MIM: 191190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG6 PubMed Links

Gene Details

Gene
PLEKHG6
Gene Name
pleckstrin homology and RhoGEF domain containing G6
There are no transcripts associated with this gene.

Gene
TNFRSF1A
Gene Name
TNF receptor superfamily member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001065.3 1290 Silent Mutation CTC,CTT L329L NP_001056.1

View Full Product Details