Product Details
- SNP ID
-
rs138291229
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120347484 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGCCGCTGCCGCCGCTGCCGCC[A/G]CCATTGGTCCGTAGGCAGTGAGAGG
- Phenotype
-
MIM: 603328
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MSI1
PubMed Links
Gene Details
- Gene
- MSI1
- Gene Name
- musashi RNA binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002442.3 |
1581 |
Missense Mutation |
GCG,GTG |
A274V |
NP_002433.1 |
XM_006719403.3 |
1581 |
Missense Mutation |
GCG,GTG |
A274V |
XP_006719466.1 |
XM_006719404.3 |
1581 |
Missense Mutation |
GCG,GTG |
A136V |
XP_006719467.1 |
XM_011538361.2 |
1581 |
Missense Mutation |
GCG,GTG |
A274V |
XP_011536663.1 |
XM_011538362.2 |
1581 |
Missense Mutation |
GCG,GTG |
A255V |
XP_011536664.1 |
XM_011538363.2 |
1581 |
Missense Mutation |
GCG,GTG |
A255V |
XP_011536665.1 |
XM_011538364.2 |
1581 |
Missense Mutation |
GCG,GTG |
A174V |
XP_011536666.1 |
XM_011538365.2 |
1581 |
Missense Mutation |
GCG,GTG |
A171V |
XP_011536667.1 |
XM_011538366.2 |
1581 |
Missense Mutation |
GCG,GTG |
A171V |
XP_011536668.1 |
XM_011538368.2 |
1581 |
Missense Mutation |
GCG,GTG |
A136V |
XP_011536670.1 |
XM_011538370.2 |
1581 |
Missense Mutation |
GCG,GTG |
A136V |
XP_011536672.1 |
XM_011538371.2 |
1581 |
Missense Mutation |
GCG,GTG |
A136V |
XP_011536673.1 |
View Full Product Details