Product Details

SNP ID
rs138564535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26336590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATTTTCTCATCTAAAAGTGTTA[C/T]TTTAACTCAGAAAACATACTGGCAT
Phenotype
MIM: 600144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITPR2 PubMed Links

Gene Details

Gene
ITPR2
Gene Name
inositol 1,4,5-trisphosphate receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002223.3 11389 UTR 3 NP_002214.2
XM_017019266.1 11389 UTR 3 XP_016874755.1
XM_017019267.1 11389 Intron XP_016874756.1
XM_017019268.1 11389 Intron XP_016874757.1
XM_017019269.1 11389 Intron XP_016874758.1

View Full Product Details