Product Details

SNP ID
rs138705814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92424897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAAGATATTATTTTACATTTCT[A/G]TATTCTCCTACCCTGAGTTGATGTG
Phenotype
MIM: 616988 MIM: 616989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLLU1 PubMed Links

Gene Details

Gene
CLLU1
Gene Name
chronic lymphocytic leukemia up-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025233.2 939 Missense Mutation ATA,GTA I73V NP_001020404.1
Gene
CLLU1OS
Gene Name
chronic lymphocytic leukemia up-regulated 1 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025232.1 939 Intron NP_001020403.1
XM_017019673.1 939 Intron XP_016875162.1

View Full Product Details